Tlačiť
Clips for standard ground joints
Dodávateľ: LENZ LABORGLAS
01103514 01103529 01103519 01103524 01103545 01103599
201-1079EA 22.35 EUR
201-1079 201-1082 201-1080 201-1081 201-1083 201-1084
Clips for standard ground joints
Svorky Svorky pro kónické NZ
Nickel-chromium steel. For conical ground joint connections.
  • Quick, easy assembly and disassembly
  • Very good thermal resistance, good chemical resistance
Objednávacie informácie: Optional fixing screws are available.