Tlačiť
Round bottom flasks with standard glass ground joint
Dodávateľ: LENZ LABORGLAS
03003170 03003149 03003137 03003158 03003128
201-1367EA 103.75 EUR
201-1367 201-1365 201-1364 201-1366 201-0908
Round bottom flasks with standard glass ground joint
Baňky Baňky s kulatým dnem
DURAN® borosilicate glass 3.3, amber.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With centre neck NS 14/23 or NS 29/32 and white matt labelling area. Coloured brown to protect light-sensitive substances.
DIN 12348
Certifikácie: DIN EN ISO 4797