Tlačiť
Adapters, receiver, Quickfit®
Dodávateľ: DWK LIFE SCIENCES
RA1/22
201-0620EA 32.29 EUR
201-0620
Adapters, receiver, Quickfit®
Komponenty pro destilaci Adaptéry destilační
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Straight delivery.