Tlačiť

Azid sodný ≥99.7%, OR, Macron Fine Chemicals™

Dodávateľ: Avantor Performance Materials
Nebezpečí

Synonymá: Sodium triazide

1953-09EA 1561.79 EUR
1953-09 1953-59
Azid sodný ≥99.7%, OR, Macron Fine Chemicals™
Azid sodný

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.

Vzorec: NaN₃
Mw: 65.01
Teplota tavenia: 275
Číslo MDL: MFCD00003536
Číslo CAS: 26628-22-8
OSN: 1687
ADR: 6.1 II
Merck Index: 13,08653

Order Now

Výsledky špecifikačného testu

For Laboratory,Research,or Manufacturing Use = 99.7 %
Assay (NaN₃) ≥99.7 %
Loss on Drying at 105°C ≤0.1 %
Typical property (not a specification): Appearance (white, crystalline
powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR