Tlačiť
Water stills, double distillation, Puridest
Dodávateľ: LAUDA
L003021 L003022 L003024 L003249 L003247 L003248 L003264 L003261 L003262
171-1312EA 7596.64 EUR
171-1312 171-1313 171-1315 171-1323 171-1321 171-1322 171-1334 171-1331 171-1332
Water stills, double distillation, Puridest
Systémy pro výrobu čisté vody Přístroje destilační
Water still (fully automatic) for double distillation, suitable for both bench and wall mounting.
  • Water still (fully automatic) for double distillation, suitable for both bench and wall mounting
  • Main switch with pilot lamp for the monitoring of both distillation stages at the front of the unit
  • Valve made of borosilicate glass 3.3 with PTFE plunger for single distillate
  • Outlet with dust guard shield made of borosilicate glass 3.3 for double distillate
  • Condenser (cooling coil) 1st stage made of stainless steel
  • Condenser (cooling coil) 2nd stage including the
More Product Information