Tlačiť
VWR®, Ultrapure Water System, U Series
Dodávateľ: VWR Collection
171-1097EA 8343.28 EUR
171-1097 171-1246 171-1098 171-1099 171-1244 171-1245 171-1467 171-1468 171-1469 171-1470 171-1471 171-1472
VWR®, Ultrapure Water System, U Series
Systémy pro výrobu čisté vody Systémy pro výrobu čisté a ultračisté vody
This 'two in one' lab water system produces ultrapure (Type I) and RO (Type III) water from tap water. Quality of ultrapure water meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user with full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
  • RFID-tagged consumables
More Product Information