Tlačiť
Gamma microcarriers, Cytodex 1
Dodávateľ: Cytiva
17-5487-01 17-5487-02 17-5487-03
17-5487-01EA 1472.86 EUR
17-5487-01 17-5487-02 17-5487-03
Gamma microcarriers, Cytodex 1
Kuličky Mikronosiče
Sterilised Cytodex 1 Gamma is a general-purpose microcarrier particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.
  • Gamma-sterilised and ready to use for quick culture start-up
  • Delivered in a convenient, single-use container system for easy transfer to various cell culture vessels
  • Supplied dry to save storage space and facilitate transportation
  • Enable culturing of anchorage-dependent cells in single-use bioreactor to eliminate the need for cleaning
More Product Information