Tlačiť
Ficoll-Paque® PLUS and Ficoll-Paque® PREMIUM Centrifugation Media, Cytiva
Dodávateľ: Cytiva
17-5442-02 17-5442-03 17-5446-02 17-5446-52 17-1440-02 17-1440-03
17-5442-02EA 386.94 EUR
17-5442-02 17-5442-03 17-5446-02 17-5446-52 17-1440-02 17-1440-03
Ficoll-Paque® PLUS and Ficoll-Paque® PREMIUM Centrifugation Media, Cytiva
Média k separaci buněk
Ficoll-Paque™ PREMIUM products are a range of sterile, ready to use density gradient media for the preparation of mononuclear cells.
  • Manufactured according to GMP and ISO 13485:2003 standards and the recommendations of the United States Pharmacopeia
  • Sterile and ready to use
  • Low levels of endotoxin (<0,12 EU/ml) secured and tested
  • Non cytotoxic
Sterile, ready to use reagents used to prepare mononuclear cell preparations from peripheral blood, bone marrow, and umbilical cord blood by density gradient centrifugation. Liquid physical state. ColorlessMore Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable