Tlačiť

Mangan 10000 mg/L Mn in 5% kyselině dusičné (from Mn) Specpure® standard pro ICP

Dodávateľ: Thermo Fisher Scientific

14413.AD 14413.AE 14413.AP
14413.ADEA 120.17 EUR
14413.AD 14413.AE 14413.AP
Mangan 10000 mg/L Mn in 5% kyselině dusičné (from Mn) Specpure® standard pro ICP
Standardy Standardy kationické Standardy - mangan

Molecular Formula: Mn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111


Certifikácie: Traceable to NIST

Order Now

ŠPECIFIKÁCIA

Environmentally Preferable
MDL čí. MFCD00011111

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR