Tlačiť

Thulium standard solution, 10000 mg/L Tm in 5% kyselině dusičné (from Tm₂O₃) Specpure® standard pro ICP

Dodávateľ: Thermo Fisher Scientific

14402.ADEA 189.87 EUR
14402.AD 14402.AE
Thulium standard solution, 10000 mg/L Tm in 5% kyselině dusičné (from Tm₂O₃) Specpure® standard pro ICP
Standardy Standardy kationické Standardy - thuluim

Molecular Formula: Tm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281


Certifikácie: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR