Tlačiť

Chromium standard dissolved in oil, 1.000 µg/g Cr in mineral oil Specpure® pro wear metal analysis

Dodávateľ: Thermo Fisher Scientific

14222.18
14222.18EA 110.55 EUR
14222.18
Chromium standard dissolved in oil, 1.000 µg/g Cr in mineral oil Specpure® pro wear metal analysis
Standardy Standardy kationické Standardy - chrom

Physical Form: Liquid
MDL No.: MFCD00010944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR