Tlačiť
Neutral detergents for cleanrooms, ClearKlens Plus VH5
Dodávateľ: DIVERSEY LEVER
7513376
141-1378EA 183.06 EUR
141-1378
Neutral detergents for cleanrooms, ClearKlens Plus VH5
Prostředky čistící Detergenty a povrchové čističe
ClearKlens Plus contains a concentrated blend of detergents and emulsifiers. Its balanced neutral formulation is particularly effective against product residues found in the pharmaceutical and cosmetic industries.
  • Available in a 30 ml single-shot bottle of concentrate for ease of dilution
  • Compatible with materials commonly found in pharmaceutical cleanrooms
  • Single-shot bottle is 0,2 µm filtered, sterile filled and double-bagged
  • Supplied with Certificate of Analysis and sterility as appropriate