Tlačiť
Anchorage sling, Titan™
Dodávateľ: HONEYWELL SAFETY PRODUCTS
1008360
139-0223EA 23.59 EUR
139-0223
Anchorage sling, Titan™
Ochrana před pádem Lana kotvící
Webbing anchorage sling for use with a personal fall limiter when working in confined spaces. For attachment to self-retracting lifelines, safety ropes and fall limiters.
  • Polyamide harness sling, 20 mm wide
  • High breaking load
  • Safe and easy to install anchorage point
Length: 0,6 m
Certifikácie: EN 795B, EN 566