Tlačiť
Powder fume hoods, NEG series
Dodávateľ: CLEANAIR BY BAKER
NP0530460 NP0530260 NP0530360 NP0530560 NP0530660 NP0531260 NP0531360 NP0531460 NP0531560 NP0531660
135-1518EA 7785.7 EUR
135-1518 135-1516 135-1517 135-1519 135-1520 135-1521 135-1522 135-1523 135-1524 135-1525
Powder fume hoods, NEG series
Digestoře Odtahy
These powder fume hoods are used wherever harmful powders or dusts are released, e.g. when weighing. The hoods guarantee the highest degree of protection for the user and the environment. The housing consists of water-tight plywood, entirely covered with a white plastic film and a hood of hardened glass.
  • The pre-filter and the main HEPA filter are efficient and easy to replace
  • Very quiet running
Certifikácie: HEPA filter: H14 conforms to EN 1822.
Informace o dodávce: Delivery, installation and commissioning prices available on request, pleaseMore Product Information