Tlačiť
Moisture barrier bags, Dri-Shield® 2000 series
Dodávateľ: DESCO
7001012 7001618
129-2645EA 70.5 EUR
129-2645 129-2646
Moisture barrier bags, Dri-Shield® 2000 series
Sáčky Sáčky skladovací
This aluminised moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area. The bags are heat sealable and suitable for vacuum packaging.
  • 0,0036 " (3.6 mil) thick bag film laminate constructed from multiple layers of aluminised polyester and polyethylene
  • Designed for moisture and electrostatic discharge (ESD) sensitive items (such as SMT)
  • <0,03 Grams Moisture Vapor Transmission Rate (MVTR); designed for dry packing of moisture sensitive devices and to protect contents from
More Product Information