Tlačiť
PIG® HazMat absorbent pillows
Dodávateľ: NEW PIG
HR7015 PIL302 PIL306 PIL307
121-0504EA 216.7 EUR
121-0504 121-0540 121-0541 121-0542
PIG® HazMat absorbent pillows
Absorpční materiály
High capacity pillows pump up the absorbing power on large spills of corrosive liquids.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulphuric acid and 30% sodium hydroxide
  • Chemical resistant pillow won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Pillow has large surface area, high capacity and fast wicking filler to quickly soak up liquids; great for adding absorbency during spill response
  • PP skin resists chemicals and tearing; reduces dust and holds in liquid,
More Product Information