Tlačiť
Safety waste systems, Nalgene®
Dodávateľ: Thermo Fisher Scientific
6379-0004 6379-0010
119-3004EA 171.04 EUR
119-3004 119-3010
Safety waste systems, Nalgene®
Láhve Láhve plastové
HDPE.
  • 10 L made from fluorinated HDPE, 4 L from HDPE
  • PP funnel includes cover with easy snap safety latch to minimise accidental spills and volatile emissions
  • Built in vent plug releases pressure during pouring and prevents emissions during use
  • Closure/adapter can also be connected to an external syringe filter
  • Removable PP screen insert/filter fits at bottom of funnel to trap large particles
A system for temporary storage of solvent, chemical, and biological waste which consists of a chemical and break resistant container and removable funnel withMore Product Information