Tlačiť
Safety Barrels
Dodávateľ: ROETZMEIER SICHERH BEHAELTER
10D 10K 10Z 25K 25Z 25ZI 50K 50Z 50ZI 10ZI 10S 10SI 200Z 200ZI 200K 10DI 100ZI 100Z 100K
119-2160EA 352.49 EUR
119-2160 119-0115 119-0119 119-0123 119-0124 119-0125 119-0126 119-0127 119-0128 119-0120 119-0117 119-0118 119-0164 119-0160 119-0158 119-0111 119-0110 119-0109 119-0108
Safety Barrels
Nádoby bezpečnostní
These safety barrels are suitable for safe and comfortable handling and storage of larger amounts of flammable liquid.
  • In-plant use of flammable liquids such as solvents
  • Explosion-proof due to flame traps (removable) and pressure control valves
  • Taps and metering devices are self-closing
  • High quality, chemically resistant stainless steel (thickness 0,8 to 1,5 mm depending on capacity) with PTFE gaskets
  • Models with self-closing tap are equipped with separate ventilation which maximises the flow rate
  • PFA content level indicator incl. a scale for volumes 10, 25
More Product Information