Tlačiť
Eyewash stations, Duo
Dodávateľ: Plum Safety
4803 4810 4802 4816
118-0096EA 90.13 EUR
118-0096 118-0097 118-0095 118-0098
Eyewash stations, Duo
Pomůcky první pomoci Sprchy na výplach očí
Wall-mounted eyewash stations, made in hard plastic, for simple and quick first-aid in case of an eye injury.
  • Easily wall-mountable
  • Ensures quick and efficient eyewash
  • Duo eye-cup for simultaneous washing of both eyes
  • Supplied with pictograms and mirror
  • Closed stations (boxes) are ideal for use in dirty and dusty areas
Eyewash station (with Eye Wash Duo):
Wall-mounted station containing 1×1000 ml Eye Wash Duo bottle, wall mounting device, pictogram and mirror. Especially for general eye washing at smaller work places.
Rinsing time: ~5 min