Tlačiť
SPINeasy RNA kit for tissue, 50 preparations, MP Biomedicals
Dodávateľ: MP Biomedicals
116543050
116543050.EA 588.02 EUR
116543050.
SPINeasy RNA kit for tissue, 50 preparations, MP Biomedicals
Činidla pro nukleové kyseliny Nucleic Acid Purification Kits and Reagents
Quick and convenient purification of total RNA from animal tissues, plant tissues and tissue cultures using spin column technology.
  • The kit allows multiple samples to be processed simultaneously without the use of toxic substances such as phenol and chloroform
  • Purified RNA is of high quality and integrity and immediately ready for RT-PCR and other downstream applications
SPINeasy RNA Kit for Tissue is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from various animal tissues plant tissues and tissue cultures.
The use of our specially formulated Lysis Buffer R and Lysing Matrix A in combination with FastPrep® instruments from MP Biomedicals enables highly efficient lysis of tissue samples within seconds. With a simple workflow, the kit allows multiple samples to be processed simultaneously without the use of toxic substances such as phenol and chloroform.
Purified RNA is of high quality and integrity and immediately ready for RT-PCR and other downstream applications.