Tlačiť
Stainless steel probe
Dodávateľ: DESCO
35123
115-3063EA 5.49 EUR
115-3063
Stainless steel probe
Čidla teplotní
Hooked stainless steel probe, 140 mm.
  • Ergonomic hex handle
  • Allows operator to better grip probe; reduces fatigue from gripping, product won't roll
  • Non-magnetic, conductive, 300 series stainless steel
  • Stainless steel won’t corrode - no problem in working with magnetised parts