Tlačiť
Centrefeed dispenser, WypAll® Reach™
Dodávateľ: KIMBERLY CLARK
6221 6241
115-2528EA 54.08 EUR
115-2528 115-2967
Centrefeed dispenser, WypAll® Reach™
Zásobníky na ubrousky
Transparent black, portable, single-sheet wiper dispenser. Ensures rolls remain clean and enclosed to help prevent transmission of food-borne bacteria. Fits service and retail centrefeed rolls (115-2527) and food and hygiene centrefeed rolls (115-2529 and 115-2531).
  • One portable centrefeed white roll or blue roll dispenser
  • Fits one white or blue roll at a time
  • Portable centrefeed roll dispenser designed for use in service, retail, and food preparation environments
  • Design ensures centrefeed rolls remain clean and enclosed to help prevent against
More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable