Tlačiť
ISO AIR cleanroom face masks with headloops
Dodávateľ: Medicom
60.701-30 60.702
114-9500EA 233.13 EUR
114-9500 114-9499
ISO AIR cleanroom face masks with headloops
Masky Roušky pro čisté prostory
ISO AIR cleanroom face masks with headloops feature an adjustable and malleable nose clip, and provide optimal wearing comfort thanks to soft and non-irritating elastic headloops.
  • Made in France
  • Made from 25% recycled polypropylene material
  • High breathability
  • ISO 3 Grade B protection
Masks feature low particle release, and are Helmke drum tested 'excellent' for filtration efficiency (BFE, PFE, VFE >99%). Headloop style masks offer increased comfort and are easy to put on and remove.
Certifikácie: The ISO AIR range is manufactured in an ISO 8More Product Information