Tlačiť
Tepelně odolné rukavice, ActivArmr® 42-445 / 42-474
Dodávateľ: Ansell
42-445/10 42-445/9 42-474/10 42-474/9 42-474/8 42-445/8
112-0288EA 25.36 EUR
112-0288 112-0289 112-3513 112-3514 112-4319 112-4302
Tepelně odolné rukavice, ActivArmr® 42-445 / 42-474
Rukavice
Superior heat resistance with complete protection for hand and wrist. Non-woven felt insulation and a full-length heat barrier designed for intermittent handling of dry objects up to 200 (42-445) or 250 °C (42-474).
  • Two-piece construction eliminates outseams for better finger comfort, dexterity and flexibility
  • Bonded nitrile coating protects against snags, punctures, abrasions and cuts
  • Provides excellent grip when handling dry or oily surfaces
  • Comfortable and flexible: sweat absorbent non-woven felt inner
  • Contains no asbestos
Recommended for: PressMore Product Information