Tlačiť
Cleanroom gloves, nitrile, Nitrilite® 93-401
Dodávateľ: Ansell
93-401/10-10.5 93-401/6-6.5 93-401/7-7.5 93-401/8-8.5 93-401/9-9.5 93-401/5-5.5
112-3478EA 487.08 EUR
112-3478 112-3479 112-3480 112-3481 112-3482 ANSE93-401/5-5.5
Cleanroom gloves, nitrile, Nitrilite® 93-401
Rukavice
These ambidextrous silicone-free gloves meet the requirements of the electronic device manufacturing and biotechnologies industries.
  • Non sterile, powder-free, textured fingertips
  • Produced in a controlled powder-free environment
  • Free of latex proteins, preventing the risk of allergies
  • Excellent puncture and chemical resistance
  • Optimal electrical properties
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Purity: <400 particles/cm²
Certifikácie: EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 VIRUS; Cat. III; AQL 1,5
Balenie: 50More Product Information