Tlačiť
Nitrile gloves, TouchNTuff® 92-500
Dodávateľ: Ansell
92-500/8.5-9 92-500/7.5-8 92-500/6.5-7 92-500/9.5-10
112-0995EA 52.87 EUR
112-0995 112-0994 112-0993 112-0996
Nitrile gloves, TouchNTuff® 92-500
Rukavice
These nitrile gloves offer up to four times the puncture resistance of comparable natural latex gloves, and three times the resistance of similar neoprene gloves. They feature a proven splash resistance against hazardous chemicals.
  • Non sterile, powdered, textured fingertips
  • "Thin Nitrile Technology" formulation for easy donning and strong grip in wet or dry conditions
  • Designed for maximum freedom of movement and minimum hand fatigue
  • Contains no natural rubber proteins: Reduces the risk of Type I allergies
Suitable for chemical handling, lightMore Product Information