Tlačiť
Screwgate karabiners, Miller®, Honeywell Safety
Dodávateľ: HONEYWELL SAFETY PRODUCTS
1002833 1002837 1018960 1009276 1018969 1018977 1018963
139-0216EA 48.87 EUR
139-0216 111-1364 139-0242 111-1391 111-1440 111-1441 111-1439
Screwgate karabiners, Miller®, Honeywell Safety
Ochrana před pádem Karabiny
The anchorage connectors and karabiners are used for securing an intermediate attachment to an anchorage point.
  • Available in different shapes, sizes and materials
Informácie o príslušenstve: EN 362