Tlačiť
Rescue triangle
Dodávateľ: HONEYWELL SAFETY PRODUCTS
1004876
111-1375EA 538.75 EUR
111-1375
Rescue triangle
Pomůcky první pomoci Zařízení pro evakuaci
This triangle is suitable for short-duration rescue and evacuation operations of an uninjured casualty in a sitting position.
  • Shoulder adjustment buckles
  • Fully adjustable and therefore suitable for adults and children
  • Compact and easy to transport