Tlačiť

FlashPure Select C18 Cartridges

Dodávateľ: BUCHI
140000121 140000122 140000123 140000124 140000125 140000126 140000127 140000128 11070490 11070491 11070492
140000121.EA 445.42 EUR
140000121. 140000122. 140000123. 140000124. 140000125. 140000126. 140000127. 140000128. 11070490. 11070491. 11070492.
FlashPure Select C18 Cartridges
Kolony chromatografické
FlashPure Select cartridge filled with spherical sorbent. FlashPure cartridges offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries.

  • C18-bonded spherical silica particles with 20 to 35 µm particle size
  • Pore size: 92 to 108 Å
  • Max. pressure: 12 bar
  • Max. loading capacity: 5%

FlashPure Select reversed phase cartridges with C18-bonded silica (20 to 35 µm, end-capped) for improved resolution in complex separations. Increased sample throughput due to high loading capacity.

Informácie o príslušenstve: UseMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR