Tlačiť
HPLC columns, ZORBAX® Eclipse XDB
Dodávateľ: AGILENT HEWLETT PACKARD
930990-902 963967-902 990967-906 993967-902 993967-906 961753-902 928700-902 823750-903 820750-903 820675-112 927975-902K 928975-902K 935967-902K 961967-902K 963967-902K 981757-302K 981758-302K 981759-302K 981759-902K 990967-902K 993967-902K
HEWL930990-902EA 1164.46 EUR
HEWL930990-902 HEWL963967-902 554-1300 HEWL993967-902 554-1301 HEWL961753-902 HEWL928700-902 HEWL823750-903 HEWL820750-903 HEWL820675-112 HEWL927975-902K HEWL928975-902K HEWL935967-902K HEWL961967-902K HEWL963967-902K HEWL981757-302K HEWL981758-302K HEWL981759-302K HEWL981759-902K HEWL990967-902K HEWL993967-902K
HPLC columns, ZORBAX® Eclipse XDB
Kolony chromatografické
Good peak shape for basic, acidic and neutral compounds.
  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping